The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001237.3 | NP_001228.1 | 5 - 6 | 1308 | 82 |
GenBank mRNA | AK291931.1 | - | 5 - 6 | 1291 | 82 |
BC104783.1 | - | 5 - 6 | 1103 | 82 | |
BC104787.1 | - | 5 - 6 | 1103 | 82 | |
CR407692.1 | - | 5 - 6 | 1003 | 82 | |
X51688.1 | - | 5 - 6 | 1110 | 82 |