The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001303256.2 | NP_001290185.1 | 25 - 26 | 3424 | 63 |
NM_014941.3 | NP_055756.1 | 26 - 27 | 4290 | 63 | |
XM_011530004.1 | XP_011528306.1 | 25 - 26 | 3649 | 63 | |
XM_017028668.1 | XP_016884157.1 | 26 - 27 | 3744 | 63 | |
GenBank mRNA | AB020659.1 | - | 26 - 27 | 4204 | 63 |
AL133637.1 | - | 7 - 8 | 895 | 63 | |
BC019257.1 | - | 24 - 25 | 2951 | 63 | |
BC136782.1 | - | 26 - 27 | 3149 | 63 | |
BU733868.1 | - | 1 - 2 | 63 | ||
CR456469.1 | - | 24 - 25 | 2950 | 63 |