The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001407.2 | NP_001398.2 | 29 - 30 | 8689 | 109 |
GenBank mRNA | AB011536.1 | - | 19 - 20 | 2565 | 109 |
AF231023.1 | - | 29 - 30 | 8689 | 109 |