Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CELSR3
Entrez Gene ID
1951
Gene Name
cadherin EGF LAG seven-pass G-type receptor 3
Gene Aliases
ADGRC3, CDHF11, EGFL1, FMI1, HFMI1, MEGF2, RESDA1
Chromosome Location
Chr.3: 48636463 - 48662915 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001407.2 NP_001398.2 3 - 4 4907 54
GenBank mRNA AF231023.1 - 3 - 4 4907 54

View Full Product Details