The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001164245.1 | NP_001157717.1 | 11 - 12 | 1354 | 147 |
NM_001164246.1 | NP_001157718.1 | 11 - 12 | 1327 | 147 | |
NM_017847.5 | NP_060317.3 | 12 - 13 | 1423 | 147 | |
XM_011509660.1 | XP_011507962.1 | 12 - 13 | 1422 | 147 | |
GenBank mRNA | AK000171.1 | - | 2 - 3 | 210 | 147 |
AK000512.1 | - | 12 - 13 | 1344 | 147 | |
AK026086.1 | - | 10 - 11 | 993 | 147 | |
AK298107.1 | - | 11 - 12 | 1263 | 147 | |
AY854248.1 | - | 12 - 13 | 1316 | 147 | |
BC003397.1 | - | 12 - 13 | 1256 | 147 | |
DQ323046.1 | - | 11 - 12 | 1187 | 147 | |
DQ529299.1 | - | 10 - 11 | 1118 | 147 |