Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TRPC7
Entrez Gene ID
57113
Gene Name
transient receptor potential cation channel subfamily C member 7
Gene Aliases
TRP7
Chromosome Location
Chr.5: 136213311 - 136365476 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001167577.1 NP_001161049.1 3 - 4 1166 133
NM_020389.2 NP_065122.1 4 - 5 1349 133
GenBank mRNA AJ272034.1 - 3 - 4 1127 133
AJ421783.1 - 3 - 4 1127 133
AJ549088.1 - 2 - 3 944 133
BC128185.1 - 3 - 4 1127 133

View Full Product Details