The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014306.4 | NP_055121.1 | 4 - 5 | 471 | 87 |
GenBank mRNA | AF155658.1 | - | 4 - 5 | 419 | 87 |
AF161466.1 | - | 4 - 5 | 430 | 87 | |
AK297908.1 | - | - | 431 | 87 | |
AK301945.1 | - | 4 - 5 | 471 | 87 | |
AK312503.1 | - | 4 - 5 | 431 | 87 | |
AL050255.1 | - | 4 - 5 | 415 | 87 | |
BC000151.1 | - | 4 - 5 | 409 | 87 | |
BC002970.1 | - | 4 - 5 | 409 | 87 | |
BC010308.1 | - | 4 - 5 | 421 | 87 | |
BC016707.1 | - | 4 - 5 | 436 | 87 | |
CR456450.1 | - | 4 - 5 | 386 | 87 | |
CR457175.1 | - | 4 - 5 | 340 | 87 | |
DC396330.1 | - | 4 - 5 | 473 | 87 |