The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014306.4 | NP_055121.1 | 8 - 9 | 1126 | 116 |
GenBank mRNA | AF155658.1 | - | 8 - 9 | 1074 | 116 |
AF161466.1 | - | 8 - 9 | 1085 | 116 | |
AK297826.1 | - | 8 - 9 | 1003 | 116 | |
AK312503.1 | - | 8 - 9 | 1086 | 116 | |
AL050255.1 | - | 8 - 9 | 1070 | 116 | |
AL137272.1 | - | 1 - 2 | 72 | 116 | |
BC000151.1 | - | 8 - 9 | 1064 | 116 | |
BC002970.1 | - | 8 - 9 | 1064 | 116 | |
BC010308.1 | - | 8 - 9 | 1076 | 116 | |
BC016707.1 | - | 8 - 9 | 1091 | 116 | |
CR456450.1 | - | 8 - 9 | 1041 | 116 | |
CR457175.1 | - | 8 - 9 | 995 | 116 |