The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001122870.2 | NP_001116342.1 | 8 - 9 | 902 | 56 |
NM_144641.3 | NP_653242.3 | 9 - 10 | 1268 | 56 | |
XM_005264880.3 | XP_005264937.1 | 8 - 9 | 1081 | 56 | |
XM_005264881.1 | XP_005264938.1 | 9 - 10 | 1157 | 56 | |
GenBank mRNA | AB474372.1 | - | 9 - 10 | 1236 | 56 |
AK096681.1 | - | 7 - 8 | 1196 | 56 | |
AK129647.1 | - | 3 - 4 | 533 | 56 | |
AL832484.1 | - | 7 - 8 | 1176 | 56 | |
BC009644.1 | - | 8 - 9 | 863 | 56 |