Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
STAG1
Entrez Gene ID
10274
Gene Name
stromal antigen 1
Gene Aliases
SA1, SCC3A
Chromosome Location
Chr.3: 136336235 - 136752412 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_005862.2 NP_005853.2 15 - 16 1839 116
XM_011512329.2 XP_011510631.1 17 - 18 2043 116
XM_011512331.2 XP_011510633.1 16 - 17 1973 116
XM_017005523.1 XP_016861012.1 14 - 15 1717 116
XM_017005524.1 XP_016861013.1 15 - 16 1913 116
XM_017005525.1 XP_016861014.1 3 - 4 411 116
GenBank mRNA AB210023.1 - 15 - 16 1804 116
AK309934.1 - 16 - 17 1939 116
BC064699.1 - 15 - 16 1814 116
Z75330.1 - 15 - 16 1947 116

View Full Product Details