The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001246.3 | NP_001237.1 | 7 - 8 | 1201 | 62 |
GenBank mRNA | U91510.1 | - | 7 - 8 | 1168 | 62 |