The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001105562.2 | NP_001099032.1 | 24 - 25 | 4017 | 93 |
NM_006048.4 | NP_006039.2 | 23 - 24 | 3630 | 93 | |
XM_005263422.1 | XP_005263479.1 | 25 - 26 | 3994 | 93 | |
XM_011540488.2 | XP_011538790.1 | 24 - 25 | 3293 | 93 | |
XM_017000035.1 | XP_016855524.1 | 22 - 23 | 3080 | 93 | |
XM_017000036.1 | XP_016855525.1 | 24 - 25 | 3294 | 93 | |
GenBank mRNA | AB014584.3 | - | 23 - 24 | 3521 | 93 |
AB028839.1 | - | 23 - 24 | 3638 | 93 | |
AF043117.1 | - | 24 - 25 | 3414 | 93 | |
AF331520.1 | - | 23 - 24 | 2989 | 93 | |
AK292444.1 | - | 23 - 24 | 3112 | 93 | |
AK304484.1 | - | 21 - 22 | 2617 | 93 | |
BC093696.1 | - | 23 - 24 | 3020 | 93 | |
BC143460.1 | - | 22 - 23 | 2905 | 93 | |
JF289274.1 | - | 25 - 26 | 3482 | 93 |