Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TALDO1
Entrez Gene ID
6888
Gene Name
transaldolase 1
Gene Aliases
TAL, TAL-H, TALDOR, TALH
Chromosome Location
Chr.11: 747432 - 765024 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_006755.1 NP_006746.1 2 - 3 273 125
GenBank mRNA AK295537.1 - 2 - 3 241 125
AK313427.1 - 2 - 3 241 125
BC010103.2 - 2 - 3 278 125
BC018847.2 - 2 - 3 221 125
L19437.2 - 2 - 3 273 125

View Full Product Details