The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004102.4 | NP_004093.1 | 1 - 2 | 347 | 64 |
XM_011541007.2 | XP_011539309.1 | 1 - 2 | 677 | 64 | |
GenBank mRNA | AK314122.1 | - | 1 - 2 | 73 | 64 |
BC007021.1 | - | 1 - 2 | 135 | 64 | |
BP194697.1 | - | 1 - 2 | 65 | ||
BT006727.1 | - | 1 - 2 | 73 | 64 | |
CN283506.1 | - | 1 - 2 | 126 | 64 | |
CR456867.1 | - | 1 - 2 | 73 | 64 | |
X56549.1 | - | 1 - 2 | 118 | 64 | |
Y10255.1 | - | 1 - 2 | 77 | 64 |