Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SLC6A3
Entrez Gene ID
6531
Gene Name
solute carrier family 6 member 3
Gene Aliases
DAT, DAT1, PKDYS
Chromosome Location
Chr.5: 1392790 - 1445428 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001044.4 NP_001035.1 13 - 14 1888 113
GenBank mRNA BC132977.1 - 12 - 13 1813 113
BC133003.1 - 12 - 13 1813 113
EF174603.1 - 13 - 14 1890 113
L24178.1 - 12 - 13 1781 113
M95167.1 - 13 - 14 1890 113
S44626.1 - 12 - 13 1787 113
S46955.1 - 13 - 14 1863 113

View Full Product Details