Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SLC6A3
Entrez Gene ID
6531
Gene Name
solute carrier family 6 member 3
Gene Aliases
DAT, DAT1, PKDYS
Chromosome Location
Chr.5: 1392790 - 1445428 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001044.4 NP_001035.1 5 - 6 914 75
GenBank mRNA BC132977.1 - 4 - 5 839 75
BC133003.1 - 4 - 5 839 75
EF174603.1 - 5 - 6 916 75
L24178.1 - 4 - 5 807 75
M95167.1 - 5 - 6 916 75
S44626.1 - 4 - 5 813 75
S46955.1 - 5 - 6 889 75

View Full Product Details