The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001243351.1 | NP_001230280.1 | 1 - 2 | 81 | 117 |
NM_016118.4 | NP_057202.3 | 1 - 2 | 81 | 117 | |
XM_017012304.1 | XP_016867793.1 | 1 - 2 | 143 | 117 | |
XM_017012305.1 | XP_016867794.1 | 1 - 2 | 143 | 117 | |
XM_017012307.1 | XP_016867796.1 | 1 - 2 | 143 | 117 | |
XM_017012308.1 | XP_016867797.1 | 1 - 2 | 143 | 117 | |
XM_017012311.1 | XP_016867800.1 | 1 - 2 | 46 | 117 | |
GenBank mRNA | AF300717.1 | - | 1 - 2 | 70 | 117 |
DB048887.1 | - | 1 - 2 | 81 | 117 |