The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001243351.1 | NP_001230280.1 | 8 - 9 | 888 | 67 |
NM_016118.4 | NP_057202.3 | 8 - 9 | 888 | 67 | |
XM_005250011.2 | XP_005250068.1 | 8 - 9 | 953 | 67 | |
XM_017012304.1 | XP_016867793.1 | 8 - 9 | 947 | 67 | |
XM_017012305.1 | XP_016867794.1 | 8 - 9 | 947 | 67 | |
XM_017012306.1 | XP_016867795.1 | 8 - 9 | 969 | 67 | |
XM_017012307.1 | XP_016867796.1 | 8 - 9 | 950 | 67 | |
XM_017012308.1 | XP_016867797.1 | 8 - 9 | 947 | 67 | |
XM_017012309.1 | XP_016867798.1 | 8 - 9 | 953 | 67 | |
XM_017012310.1 | XP_016867799.1 | 9 - 10 | 1050 | 67 | |
XM_017012311.1 | XP_016867800.1 | 9 - 10 | 954 | 67 | |
XM_017012312.1 | XP_016867801.1 | 9 - 10 | 923 | 67 | |
XM_017012313.1 | XP_016867802.1 | 9 - 10 | 1050 | 67 | |
XM_017012314.1 | XP_016867803.1 | 9 - 10 | 926 | 67 | |
GenBank mRNA | AF155099.1 | - | 8 - 9 | 820 | 67 |
AF300717.1 | - | 8 - 9 | 877 | 67 | |
AF459743.1 | - | 8 - 9 | 836 | 67 | |
AY129295.1 | - | 8 - 9 | 836 | 67 | |
BC046354.2 | - | 7 - 8 | 804 | 67 | |
BG105733.1 | - | 1 - 2 | 120 | 67 |