The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_007039.3 | NP_008970.2 | 18 - 19 | 3727 | 84 |
XM_005267287.2 | XP_005267344.1 | 18 - 19 | 3776 | 84 | |
XM_006720011.3 | XP_006720074.1 | 17 - 18 | 3569 | 84 | |
XM_011536367.2 | XP_011534669.1 | 18 - 19 | 6895 | 84 | |
XM_011536368.2 | XP_011534670.1 | 16 - 17 | 3139 | 84 | |
XM_011536369.2 | XP_011534671.1 | 18 - 19 | 3721 | 84 | |
XM_017020938.1 | XP_016876427.1 | 17 - 18 | 3678 | 84 | |
XM_017020939.1 | XP_016876428.1 | 17 - 18 | 3267 | 84 | |
GenBank mRNA | AI800682.1 | - | 1 - 2 | 84 | |
BC143847.1 | - | 16 - 17 | 3697 | 84 | |
X79510.1 | - | 18 - 19 | 3728 | 84 |