Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SNRNP40
Entrez Gene ID
9410
Gene Name
small nuclear ribonucleoprotein U5 subunit 40
Gene Aliases
40K, HPRP8BP, PRP8BP, PRPF8BP, SPF38, WDR57
Chromosome Location
Chr.1: 31259568 - 31296797 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_004814.2 NP_004805.2 6 - 7 825 146
GenBank mRNA AF090988.1 - 6 - 7 794 146
AK074516.1 - 6 - 7 808 146
AK123103.1 - 2 - 3 1235 146
AK298823.1 - 6 - 7 843 146
AK313800.1 - 6 - 7 790 146
BC001494.2 - 6 - 7 801 146
EF011621.1 - 6 - 7 800 146

View Full Product Details