Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SNRNP40
Entrez Gene ID
9410
Gene Name
small nuclear ribonucleoprotein U5 subunit 40
Gene Aliases
40K, HPRP8BP, PRP8BP, PRPF8BP, SPF38, WDR57
Chromosome Location
Chr.1: 31259568 - 31296797 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_004814.2 NP_004805.2 8 - 9 963 96
GenBank mRNA AF090988.1 - 8 - 9 932 96
AK074516.1 - 8 - 9 946 96
AK123103.1 - 4 - 5 1373 96
AK298823.1 - 8 - 9 981 96
AK313800.1 - 8 - 9 928 96
AL157420.1 - 2 - 3 568 96
BC001494.2 - 8 - 9 939 96
EF011621.1 - 8 - 9 938 96

View Full Product Details