The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001048166.1 | NP_001041631.1 | 5 - 6 | 605 | 78 |
NM_001282936.1 | NP_001269865.1 | 6 - 7 | 814 | 78 | |
NM_001282937.1 | NP_001269866.1 | 6 - 7 | 814 | 78 | |
NM_001282938.1 | NP_001269867.1 | 6 - 7 | 814 | 78 | |
NM_001282939.1 | NP_001269868.1 | 6 - 7 | 679 | 78 | |
NM_003035.2 | NP_003026.2 | 5 - 6 | 605 | 78 | |
XM_006710834.3 | XP_006710897.1 | 6 - 7 | 679 | 78 | |
XM_011541991.2 | XP_011540293.1 | 6 - 7 | 1345 | 78 | |
XM_011541992.2 | XP_011540294.1 | 6 - 7 | 1328 | 78 | |
XM_011541994.2 | XP_011540296.1 | 5 - 6 | 605 | 78 | |
XM_011541996.2 | XP_011540298.1 | 5 - 6 | 605 | 78 | |
XM_011541998.2 | XP_011540300.1 | 5 - 6 | 605 | 78 | |
XM_011542001.2 | XP_011540303.1 | 5 - 6 | 605 | 78 | |
XM_017002123.1 | XP_016857612.1 | 5 - 6 | 605 | 78 | |
XM_017002125.1 | XP_016857614.1 | 5 - 6 | 605 | 78 | |
XM_017002126.1 | XP_016857615.1 | 5 - 6 | 605 | 78 | |
XM_017002127.1 | XP_016857616.1 | 5 - 6 | 605 | 78 | |
GenBank mRNA | AK307519.1 | - | 5 - 6 | 567 | 78 |
BC053615.1 | - | 6 - 7 | 647 | 78 | |
BC126223.1 | - | 5 - 6 | 681 | 78 | |
BC144089.1 | - | 5 - 6 | 681 | 78 | |
CR749851.1 | - | 5 - 6 | 581 | 78 | |
M74558.1 | - | 6 - 7 | 830 | 78 |