Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
STIL
Entrez Gene ID
6491
Gene Name
SCL/TAL1 interrupting locus
Gene Aliases
MCPH7, SIL
Chromosome Location
Chr.1: 47250139 - 47314787 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001048166.1 NP_001041631.1 7 - 8 945 124
NM_001282936.1 NP_001269865.1 8 - 9 1154 124
NM_001282937.1 NP_001269866.1 8 - 9 1154 124
NM_001282938.1 NP_001269867.1 9 - 10 1013 124
NM_001282939.1 NP_001269868.1 9 - 10 878 124
NM_003035.2 NP_003026.2 7 - 8 945 124
XM_006710834.3 XP_006710897.1 8 - 9 1019 124
XM_011541991.2 XP_011540293.1 8 - 9 1685 124
XM_011541992.2 XP_011540294.1 8 - 9 1668 124
XM_011541994.2 XP_011540296.1 7 - 8 945 124
XM_011541996.2 XP_011540298.1 8 - 9 804 124
XM_011541998.2 XP_011540300.1 8 - 9 804 124
XM_011542001.2 XP_011540303.1 7 - 8 945 124
XM_017002123.1 XP_016857612.1 8 - 9 804 124
XM_017002124.1 XP_016857613.1 3 - 4 221 124
XM_017002125.1 XP_016857614.1 7 - 8 945 124
XM_017002126.1 XP_016857615.1 7 - 8 945 124
XM_017002127.1 XP_016857616.1 7 - 8 945 124
GenBank mRNA AK307519.1 - 7 - 8 907 124
BC053615.1 - 9 - 10 846 124
BC126223.1 - 7 - 8 1021 124
BC144089.1 - 7 - 8 1021 124
CR749851.1 - 7 - 8 921 124
M74558.1 - 8 - 9 1170 124

View Full Product Details