Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RUNX1T1
Entrez Gene ID
862
Gene Name
RUNX1 translocation partner 1
Gene Aliases
AML1-MTG8, AML1T1, CBFA2T1, CDR, ETO, MTG8, ZMYND2
Chromosome Location
Chr.8: 91954967 - 92103365 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001198625.1 NP_001185554.1 8 - 9 1238 92
NM_001198626.1 NP_001185555.1 6 - 7 1096 92
NM_001198627.1 NP_001185556.1 7 - 8 1156 92
NM_001198628.1 NP_001185557.1 5 - 6 753 92
NM_001198629.1 NP_001185558.1 5 - 6 879 92
NM_001198630.1 NP_001185559.1 6 - 7 939 92
NM_001198631.1 NP_001185560.1 5 - 6 952 92
NM_001198632.1 NP_001185561.1 5 - 6 870 92
NM_001198633.1 NP_001185562.1 4 - 5 841 92
NM_001198634.1 NP_001185563.1 4 - 5 779 92
NM_001198679.1 NP_001185608.1 5 - 6 923 92
NM_004349.3 NP_004340.1 4 - 5 889 92
NM_175634.2 NP_783552.1 5 - 6 1014 92
NM_175635.2 NP_783553.1 4 - 5 706 92
NM_175636.2 NP_783554.1 3 - 4 788 92
XM_006716676.3 XP_006716739.1 6 - 7 823 92
XM_011517351.2 XP_011515653.1 6 - 7 868 92
XM_011517352.2 XP_011515654.1 6 - 7 1108 92
XM_011517353.2 XP_011515655.1 5 - 6 816 92
XM_017013930.1 XP_016869419.1 5 - 6 1124 92
XM_017013931.1 XP_016869420.1 4 - 5 646 92
XM_017013932.1 XP_016869421.1 6 - 7 1011 92
XM_017013933.1 XP_016869422.1 7 - 8 1075 92
XM_017013934.1 XP_016869423.1 7 - 8 1183 92
XM_017013935.1 XP_016869424.1 6 - 7 988 92
XM_017013936.1 XP_016869425.1 5 - 6 1082 92
XM_017013937.1 XP_016869426.1 6 - 7 908 92
XM_017013938.1 XP_016869427.1 5 - 6 772 92
GenBank mRNA AK297616.1 - 4 - 5 603 92
BC005850.1 - 4 - 5 668 92
BC067078.1 - 4 - 5 659 92
BT009871.1 - 3 - 4 448 92
CR456792.1 - 3 - 4 478 92
D14289.1 - 3 - 4 745 92
D14821.1 - 4 - 5 643 92
D43638.1 - 4 - 5 889 92
KF946505.1 - 4 - 5 507 92
KF946507.1 - 4 - 5 507 92
KF946508.1 - 4 - 5 507 92
KF946510.1 - 4 - 5 507 92
KF946511.1 - 4 - 5 493 92
KF946512.1 - 5 - 6 1797 92
KF946513.1 - 4 - 5 2177 92
KF946514.1 - 4 - 5 493 92
KF946515.1 - 3 - 4 733 92
KF946516.1 - 3 - 4 733 92
KF946517.1 - 3 - 4 733 92
KF946518.1 - 3 - 4 733 92
X79990.1 - 3 - 4 471 92

View Full Product Details