Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
KLHL28
Entrez Gene ID
54813
Gene Name
kelch like family member 28
Gene Aliases
BTBD5
Chromosome Location
Chr.14: 44924310 - 44961978 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001308112.1 NP_001295041.1 4 - 5 1800 82
NM_017658.4 NP_060128.2 4 - 5 1685 82
XM_005267770.3 XP_005267827.1 4 - 5 1925 82
XM_011536847.2 XP_011535149.1 5 - 6 1774 82
GenBank mRNA AK289855.1 - 4 - 5 1674 82
BC121009.1 - 4 - 5 1597 82
BC121010.1 - 4 - 5 1597 82
CA433205.1 - 1 - 2 82

View Full Product Details