Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PRKCG
Entrez Gene ID
5582
Gene Name
protein kinase C gamma
Gene Aliases
PKC-gamma, PKCC, PKCG, SCA14
Chromosome Location
Chr.19: 53882184 - 53907647 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001316329.1 NP_001303258.1 17 - 18 2215 136
NM_002739.4 NP_002730.1 17 - 18 2215 136
GenBank mRNA AF345987.1 - 17 - 18 2099 136
AK296412.1 - 17 - 18 2223 136
AK296434.1 - 17 - 18 2202 136
AK303741.1 - 14 - 15 1514 136
AK312302.1 - 17 - 18 2177 136
BC047876.1 - 17 - 18 2090 136
Z15114.1 - 13 - 14 1423 136

View Full Product Details