The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001316329.1 | NP_001303258.1 | 5 - 6 | 839 | 98 |
NM_002739.4 | NP_002730.1 | 5 - 6 | 839 | 98 | |
GenBank mRNA | AB209671.1 | - | 5 - 6 | 558 | 98 |
AF345987.1 | - | 5 - 6 | 723 | 98 | |
AK296412.1 | - | 5 - 6 | 847 | 98 | |
AK296434.1 | - | 5 - 6 | 826 | 98 | |
AK302958.1 | - | 5 - 6 | 824 | 98 | |
AK303741.1 | - | 3 - 4 | 246 | 98 | |
AK312302.1 | - | 5 - 6 | 801 | 98 | |
BC047876.1 | - | 5 - 6 | 714 | 98 | |
M13977.1 | - | 5 - 6 | 723 | 98 | |
Z15114.1 | - | 1 - 2 | 47 | 98 |