The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_080658.1 | NP_542389.1 | 7 - 8 | 918 | 86 |
XM_017018549.1 | XP_016874038.1 | 8 - 9 | 1109 | 86 | |
XM_017018550.1 | XP_016874039.1 | 8 - 9 | 1143 | 86 | |
XM_017018551.1 | XP_016874040.1 | 8 - 9 | 1064 | 86 | |
GenBank mRNA | AY040761.1 | - | 7 - 8 | 898 | 86 |
AY169233.1 | - | 7 - 8 | 918 | 86 | |
BC008689.1 | - | 7 - 8 | 918 | 86 |