Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MARK2
Entrez Gene ID
2011
Gene Name
microtubule affinity regulating kinase 2
Gene Aliases
EMK-1, EMK1, PAR-1, Par-1b, Par1b
Chromosome Location
Chr.11: 63838928 - 63911020 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001039469.2 NP_001034558.2 2 - 3 811 68
NM_001163296.1 NP_001156768.1 2 - 3 811 68
NM_001163297.1 NP_001156769.1 2 - 3 811 68
NM_004954.4 NP_004945.4 2 - 3 811 68
NM_017490.3 NP_059672.2 2 - 3 628 68
XM_011544789.1 XP_011543091.1 3 - 4 702 68
XM_011544790.1 XP_011543092.1 3 - 4 702 68
XM_011544791.1 XP_011543093.1 3 - 4 702 68
XM_011544792.1 XP_011543094.1 3 - 4 702 68
XM_011544793.1 XP_011543095.1 3 - 4 702 68
XM_011544794.1 XP_011543096.1 3 - 4 702 68
XM_011544795.1 XP_011543097.1 3 - 4 702 68
XM_011544796.1 XP_011543098.1 3 - 4 702 68
XM_011544797.1 XP_011543099.1 3 - 4 702 68
XM_011544798.1 XP_011543100.1 3 - 4 702 68
XM_017017310.1 XP_016872799.1 2 - 3 784 68
XM_017017311.1 XP_016872800.1 3 - 4 702 68
GenBank mRNA AB188493.1 - 2 - 3 815 68
AB271694.1 - 1 - 2 133 68
AF387638.1 - 1 - 2 151 68
AK225766.1 - 2 - 3 646 68
AK290339.1 - 2 - 3 596 68
AK307858.1 - 3 - 4 447 68
BC008771.2 - 2 - 3 203 68
BC084540.1 - 2 - 3 270 68
BT007342.1 - 1 - 2 133 68
BX383836.2 - 2 - 3 792 68
X97630.1 - 2 - 3 540 68

View Full Product Details