Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MARK2
Entrez Gene ID
2011
Gene Name
microtubule affinity regulating kinase 2
Gene Aliases
EMK-1, EMK1, PAR-1, Par-1b, Par1b
Chromosome Location
Chr.11: 63838928 - 63911020 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001039469.2 NP_001034558.2 5 - 6 983 65
NM_001163296.1 NP_001156768.1 5 - 6 983 65
NM_001163297.1 NP_001156769.1 5 - 6 983 65
NM_004954.4 NP_004945.4 5 - 6 983 65
NM_017490.3 NP_059672.2 5 - 6 800 65
XM_011544789.1 XP_011543091.1 6 - 7 874 65
XM_011544790.1 XP_011543092.1 6 - 7 874 65
XM_011544791.1 XP_011543093.1 6 - 7 874 65
XM_011544792.1 XP_011543094.1 6 - 7 874 65
XM_011544793.1 XP_011543095.1 6 - 7 874 65
XM_011544794.1 XP_011543096.1 6 - 7 874 65
XM_011544795.1 XP_011543097.1 6 - 7 874 65
XM_011544796.1 XP_011543098.1 6 - 7 874 65
XM_011544797.1 XP_011543099.1 6 - 7 874 65
XM_011544798.1 XP_011543100.1 6 - 7 874 65
XM_017017310.1 XP_016872799.1 5 - 6 956 65
XM_017017311.1 XP_016872800.1 6 - 7 874 65
GenBank mRNA AB188493.1 - 5 - 6 987 65
AB271694.1 - 4 - 5 305 65
AF387638.1 - 4 - 5 323 65
AK225766.1 - 5 - 6 818 65
AK290339.1 - 5 - 6 768 65
BC008771.2 - 5 - 6 375 65
BC084540.1 - 5 - 6 442 65
BT007342.1 - 4 - 5 305 65
X97630.1 - 5 - 6 712 65

View Full Product Details