The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001136528.1 | NP_001130000.1 | 4 - 5 | 928 | 101 |
NM_001136530.1 | NP_001130002.1 | 4 - 5 | 858 | 101 | |
NM_006216.3 | NP_006207.1 | 4 - 5 | 928 | 101 | |
NR_073116.1 | 5 - 6 | 1346 | 101 | ||
XM_005246641.2 | XP_005246698.1 | 4 - 5 | 1017 | 101 | |
XM_017004329.1 | XP_016859818.1 | 4 - 5 | 909 | 101 | |
XM_017004330.1 | XP_016859819.1 | 4 - 5 | 746 | 101 | |
XM_017004331.1 | XP_016859820.1 | 4 - 5 | 909 | 101 | |
XM_017004332.1 | XP_016859821.1 | 4 - 5 | 746 | 101 | |
GenBank mRNA | A03911.1 | - | 3 - 4 | 709 | 101 |
AK225823.1 | - | 4 - 5 | 896 | 101 | |
AK295564.1 | - | 4 - 5 | 826 | 101 | |
AK297588.1 | - | 4 - 5 | 733 | 101 | |
AK312499.1 | - | 4 - 5 | 942 | 101 | |
BC015663.2 | - | 4 - 5 | 885 | 101 | |
BC042628.1 | - | 5 - 6 | 1345 | 101 | |
M17783.1 | - | 3 - 4 | 685 | 101 |