The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001136528.1 | NP_001130000.1 | 6 - 7 | 1234 | 136 |
NM_001136530.1 | NP_001130002.1 | 6 - 7 | 1164 | 136 | |
NR_073116.1 | 7 - 8 | 1652 | 136 | ||
XM_017004331.1 | XP_016859820.1 | 6 - 7 | 1215 | 136 | |
XM_017004332.1 | XP_016859821.1 | 6 - 7 | 1052 | 136 | |
GenBank mRNA | AK225823.1 | - | 6 - 7 | 1202 | 136 |
AK295564.1 | - | 6 - 7 | 1132 | 136 | |
AK297588.1 | - | 6 - 7 | 1039 | 136 | |
BC042628.1 | - | 7 - 8 | 1651 | 136 | |
M17783.1 | - | 5 - 6 | 991 | 136 |