Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ZFYVE1
Entrez Gene ID
53349
Gene Name
zinc finger FYVE-type containing 1
Gene Aliases
DFCP1, PPP1R172, SR3, TAFF1, ZNFN2A1
Chromosome Location
Chr.14: 72969445 - 73027212 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001281734.1 NP_001268663.1 11 - 12 2779 58
NM_001281735.1 NP_001268664.1 8 - 9 957 58
NM_021260.3 NP_067083.1 11 - 12 2821 58
NM_178441.2 NP_848535.1 8 - 9 962 58
XM_017021373.1 XP_016876862.1 9 - 10 1486 58
GenBank mRNA AB046809.1 - 11 - 12 2646 58
AF251025.2 - 10 - 11 2526 58
AF311602.1 - 8 - 9 962 58
AF318319.1 - 8 - 9 990 58
AI073571.1 - 1 - 2 58
AJ310569.1 - 11 - 12 2672 58
AK027399.1 - 11 - 12 2715 58
AK098542.1 - 8 - 9 945 58
BC014902.1 - 11 - 12 2698 58
BC053520.1 - 11 - 12 2749 58
BU740588.1 - 1 - 2 58

View Full Product Details