Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ZFYVE1
Entrez Gene ID
53349
Gene Name
zinc finger FYVE-type containing 1
Gene Aliases
DFCP1, PPP1R172, SR3, TAFF1, ZNFN2A1
Chromosome Location
Chr.14: 72969445 - 73027212 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001281734.1 NP_001268663.1 5 - 6 2032 76
NM_001281735.1 NP_001268664.1 2 - 3 168 76
NM_021260.3 NP_067083.1 5 - 6 2032 76
NM_178441.2 NP_848535.1 2 - 3 173 76
XM_017021373.1 XP_016876862.1 3 - 4 697 76
GenBank mRNA AB046809.1 - 5 - 6 1857 76
AF251025.2 - 4 - 5 1737 76
AF311602.1 - 2 - 3 173 76
AF318319.1 - 2 - 3 200 76
AJ310569.1 - 5 - 6 1883 76
AK027399.1 - 5 - 6 1926 76
AK098542.1 - 2 - 3 156 76
BC014902.1 - 5 - 6 1951 76
BC053520.1 - 5 - 6 1960 76

View Full Product Details