The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_177401.4 | NP_796375.3 | 6 - 7 | 1377 | 92 |
XM_005259671.3 | XP_005259728.1 | 7 - 8 | 1506 | 92 | |
XM_005259672.3 | XP_005259729.1 | 7 - 8 | 1504 | 92 | |
GenBank mRNA | BC094778.1 | - | 5 - 6 | 1271 | 92 |