Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TMEM259
Entrez Gene ID
91304
Gene Name
transmembrane protein 259
Gene Aliases
ASBABP1, C19orf6, MBRL, MEMBRALIN, R32184_3
Chromosome Location
Chr.19: 1009651 - 1021169 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001033026.1 NP_001028198.1 4 - 5 864 61
NM_033420.3 NP_219488.1 4 - 5 864 61
XM_005259675.2 XP_005259732.1 4 - 5 909 61
XM_005259676.2 XP_005259733.1 4 - 5 891 61
XM_005259677.2 XP_005259734.1 4 - 5 909 61
XM_005259678.2 XP_005259735.1 4 - 5 909 61
XM_017027457.1 XP_016882946.1 4 - 5 909 61
GenBank mRNA AK095756.1 - 4 - 5 864 61
BC008957.2 - 4 - 5 837 61
BQ895919.1 - 4 - 5 635 61
DQ005958.1 - 4 - 5 846 61

View Full Product Details