Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TMEM259
Entrez Gene ID
91304
Gene Name
transmembrane protein 259
Gene Aliases
ASBABP1, C19orf6, MBRL, MEMBRALIN, R32184_3
Chromosome Location
Chr.19: 1009651 - 1021169 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001033026.1 NP_001028198.1 9 - 10 1360 90
XM_005259675.2 XP_005259732.1 9 - 10 1405 90
XM_005259676.2 XP_005259733.1 9 - 10 1387 90
XM_005259677.2 XP_005259734.1 9 - 10 1354 90
XM_005259678.2 XP_005259735.1 9 - 10 1405 90
XM_017027457.1 XP_016882946.1 9 - 10 1354 90
GenBank mRNA AK090400.1 - 5 - 6 425 90
DQ005958.1 - 9 - 10 1342 90

View Full Product Details