Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CAV3
Entrez Gene ID
859
Gene Name
caveolin 3
Gene Aliases
LGMD1C, LQT9, VIP-21, VIP21
Chromosome Location
Chr.3: 8733800 - 8746765 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001234.4 NP_001225.1 1 - 2 182 69
NM_033337.2 NP_203123.1 1 - 2 182 69
GenBank mRNA AF036365.1 - 1 - 2 152 69
AF043101.1 - 1 - 2 102 69
AK291892.1 - 1 - 2 69
BC069368.1 - 1 - 2 69
BC102033.1 - 1 - 2 142 69
BC102036.1 - 1 - 2 142 69
BC102037.1 - 1 - 2 142 69
Y14747.1 - 1 - 2 116 69

View Full Product Details