Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- PER3
- Entrez Gene ID
-
8863
- Gene Name
- period circadian clock 3
- Gene Aliases
-
FASPS3, GIG13
- Chromosome Location
-
Chr.1: 7784285 - 7845181 on Build GRCh38
View Full Product Details