Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PER3
Entrez Gene ID
8863
Gene Name
period circadian clock 3
Gene Aliases
FASPS3, GIG13
Chromosome Location
Chr.1: 7784285 - 7845181 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001289861.1 NP_001276790.1 8 - 9 1138 120
NM_001289862.1 NP_001276791.1 8 - 9 1138 120
NM_001289863.1 NP_001276792.1 8 - 9 1138 120
NM_001289864.1 NP_001276793.1 8 - 9 1199 120
NM_016831.2 NP_058515.1 8 - 9 1135 120
XM_005263524.3 XP_005263581.1 7 - 8 1013 120
XM_011542385.2 XP_011540687.1 7 - 8 1013 120
XM_011542390.2 XP_011540692.1 7 - 8 1013 120
XM_017002723.1 XP_016858212.1 7 - 8 1010 120
XM_017002724.1 XP_016858213.1 7 - 8 1010 120
XM_017002725.1 XP_016858214.1 7 - 8 1013 120
XM_017002726.1 XP_016858215.1 7 - 8 1010 120
XM_017002727.1 XP_016858216.1 9 - 10 1332 120
XM_017002728.1 XP_016858217.1 7 - 8 1074 120
XM_017002729.1 XP_016858218.1 5 - 6 514 120
XM_017002730.1 XP_016858219.1 7 - 8 1074 120
XM_017002731.1 XP_016858220.1 7 - 8 1074 120
XM_017002732.1 XP_016858221.1 6 - 7 1123 120
XM_017002733.1 XP_016858222.1 9 - 10 1239 120
XM_017002734.1 XP_016858223.1 10 - 11 1381 120
XM_017002735.1 XP_016858224.1 9 - 10 1239 120
XM_017002736.1 XP_016858225.1 10 - 11 1381 120
XM_017002737.1 XP_016858226.1 5 - 6 624 120
XM_017002738.1 XP_016858227.1 5 - 6 630 120
GenBank mRNA AB047686.1 - 7 - 8 1098 120
AK307760.1 - 7 - 8 1013 120
AK308303.1 - 8 - 9 1187 120
AL157954.1 - 7 - 8 1046 120
AY493418.1 - 7 - 8 942 120
BC026102.1 - 8 - 9 1157 120
BC144137.1 - 7 - 8 1103 120
BC146781.1 - 7 - 8 1098 120

View Full Product Details