Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GSK3A
Entrez Gene ID
2931
Gene Name
glycogen synthase kinase 3 alpha
Gene Aliases
-
Chromosome Location
Chr.19: 42230186 - 42243330 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_019884.2 NP_063937.2 6 - 7 1022 82
GenBank mRNA BC027984.1 - 6 - 7 1041 82
BC051865.2 - 6 - 7 1011 82
CR749620.1 - 5 - 6 2370 82
D63424.1 - 6 - 7 994 82
L40027.1 - 6 - 7 1017 82

View Full Product Details