The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001206673.1 | NP_001193602.1 | 3 - 4 | 347 | 98 |
NM_181533.3 | NP_853511.2 | 2 - 3 | 219 | 98 | |
XM_011536472.1 | XP_011534774.1 | 3 - 4 | 417 | 98 | |
XM_011536474.2 | XP_011534776.1 | 3 - 4 | 868 | 98 | |
XM_017021031.1 | XP_016876520.1 | 2 - 3 | 281 | 98 | |
GenBank mRNA | AY311396.1 | - | 3 - 4 | 347 | 98 |
AY311397.1 | - | 2 - 3 | 219 | 98 | |
AY311399.1 | - | 2 - 3 | 219 | 98 | |
BC034603.1 | - | 1 - 2 | 83 | 98 |