Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ABHD12B
Entrez Gene ID
145447
Gene Name
abhydrolase domain containing 12B
Gene Aliases
BEM46L3, C14orf29, c14_5314
Chromosome Location
Chr.14: 50872090 - 50904970 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001206673.1 NP_001193602.1 5 - 6 506 86
NM_181533.3 NP_853511.2 4 - 5 378 86
NM_181814.1 NP_861535.1 3 - 4 275 86
XM_006720052.1 XP_006720115.1 3 - 4 272 86
XM_011536472.1 XP_011534774.1 5 - 6 576 86
XM_011536474.2 XP_011534776.1 5 - 6 1027 86
XM_017021031.1 XP_016876520.1 4 - 5 440 86
GenBank mRNA AK128464.1 - 3 - 4 272 86
AY311396.1 - 5 - 6 506 86
AY311397.1 - 4 - 5 378 86
AY311398.1 - 3 - 4 275 86
BC034603.1 - 3 - 4 242 86
BC107141.1 - 3 - 4 275 86
BC107142.1 - 3 - 4 273 86

View Full Product Details