Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ABHD12B
Entrez Gene ID
145447
Gene Name
abhydrolase domain containing 12B
Gene Aliases
BEM46L3, C14orf29, c14_5314
Chromosome Location
Chr.14: 50872090 - 50904970 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001206673.1 NP_001193602.1 6 - 7 551 67
NM_181533.3 NP_853511.2 5 - 6 423 67
NM_181814.1 NP_861535.1 4 - 5 320 67
XM_006720052.1 XP_006720115.1 4 - 5 317 67
XM_011536472.1 XP_011534774.1 6 - 7 621 67
XM_011536474.2 XP_011534776.1 6 - 7 1072 67
XM_017021031.1 XP_016876520.1 5 - 6 485 67
GenBank mRNA AK128464.1 - 4 - 5 317 67
AY311396.1 - 6 - 7 551 67
AY311397.1 - 5 - 6 423 67
AY311398.1 - 4 - 5 320 67
BC034603.1 - 4 - 5 287 67
BC107141.1 - 4 - 5 320 67
BC107142.1 - 4 - 5 318 67

View Full Product Details