The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006206.4 | NP_006197.1 | 15 - 16 | 2481 | 84 |
XM_005265743.1 | XP_005265800.1 | 15 - 16 | 2265 | 84 | |
XM_006714039.3 | XP_006714102.1 | 16 - 17 | 2526 | 84 | |
XM_006714041.3 | XP_006714104.1 | 16 - 17 | 2529 | 84 | |
XM_011534385.2 | XP_011532687.1 | 15 - 16 | 2519 | 84 | |
XM_017008280.1 | XP_016863769.1 | 15 - 16 | 2696 | 84 | |
XM_017008281.1 | XP_016863770.1 | 15 - 16 | 2762 | 84 | |
XM_017008282.1 | XP_016863771.1 | 15 - 16 | 2410 | 84 | |
GenBank mRNA | AK311006.1 | - | 14 - 15 | 1968 | 84 |
AK316578.1 | - | 15 - 16 | 2283 | 84 | |
L25829.1 | - | 3 - 4 | 575 | 84 | |
M21574.1 | - | 15 - 16 | 2289 | 84 | |
M22734.1 | - | - | 2278 | 84 | |
X76079.1 | - | 3 - 4 | 575 | 84 |