Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ATXN7
Entrez Gene ID
6314
Gene Name
ataxin 7
Gene Aliases
ADCAII, OPCA3, SCA7
Chromosome Location
Chr.3: 63864557 - 64003462 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_000333.3 NP_000324.1 1 - 2 448 91
GenBank mRNA AF032105.1 - 1 - 2 448 91
AJ000517.1 - 1 - 2 456 91

View Full Product Details