The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000333.3 | NP_000324.1 | 9 - 10 | 1920 | 130 |
NM_001128149.2 | NP_001121621.2 | 5 - 6 | 1100 | 130 | |
NM_001177387.1 | NP_001170858.1 | 8 - 9 | 1477 | 130 | |
GenBank mRNA | AF032103.1 | - | 7 - 8 | 1059 | 130 |
AF032105.1 | - | 9 - 10 | 1920 | 130 | |
AJ000517.1 | - | 9 - 10 | 1928 | 130 | |
AK304062.1 | - | 5 - 6 | 1023 | 130 |