The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001102.3 | NP_001093.1 | 11 - 12 | 1542 | 68 |
NM_001130004.1 | NP_001123476.1 | 11 - 12 | 1542 | 68 | |
NM_001130005.1 | NP_001123477.1 | 11 - 12 | 1542 | 68 | |
XM_011537267.2 | XP_011535569.1 | 11 - 12 | 1334 | 68 | |
XM_011537268.2 | XP_011535570.1 | 11 - 12 | 1334 | 68 | |
XM_017021721.1 | XP_016877210.1 | 11 - 12 | 1334 | 68 | |
XM_017021723.1 | XP_016877212.1 | 11 - 12 | 1479 | 68 | |
XM_017021726.1 | XP_016877215.1 | 11 - 12 | 1653 | 68 | |
XM_017021728.1 | XP_016877217.1 | 11 - 12 | 1653 | 68 | |
GenBank mRNA | AK098203.1 | - | 3 - 4 | 275 | 68 |
AK225782.1 | - | 11 - 12 | 1439 | 68 | |
AK294317.1 | - | 5 - 6 | 745 | 68 | |
AK295099.1 | - | 11 - 12 | 1438 | 68 | |
AK298496.1 | - | 11 - 12 | 1317 | 68 | |
AK299362.1 | - | 11 - 12 | 1180 | 68 | |
BC003576.1 | - | 11 - 12 | 1474 | 68 | |
BC015766.1 | - | 11 - 12 | 1349 | 68 | |
BT007207.1 | - | 11 - 12 | 1232 | 68 | |
BX248020.1 | - | 11 - 12 | 1205 | 68 | |
DQ496098.1 | - | 11 - 12 | 1415 | 68 | |
EU716325.1 | - | 11 - 12 | 1252 | 68 | |
FJ410030.1 | - | 11 - 12 | 1252 | 68 | |
M95178.1 | - | 11 - 12 | 1343 | 68 | |
X15804.1 | - | 11 - 12 | 1430 | 68 | |
X55187.1 | - | 2 - 3 | 344 | 68 |