The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001284206.1 | NP_001271135.1 | 7 - 8 | 1162 | 67 |
NM_001284207.1 | NP_001271136.1 | 7 - 8 | 918 | 67 | |
NM_003861.2 | NP_003852.1 | 7 - 8 | 1165 | 67 | |
XM_006720297.2 | XP_006720360.1 | 8 - 9 | 1270 | 67 | |
XM_006720298.2 | XP_006720361.1 | 8 - 9 | 1267 | 67 | |
XM_006720299.3 | XP_006720362.1 | 8 - 9 | 905 | 67 | |
XM_006720300.2 | XP_006720363.1 | 8 - 9 | 926 | 67 | |
XM_011537278.1 | XP_011535580.1 | 9 - 10 | 1539 | 67 | |
XM_011537279.2 | XP_011535581.1 | 8 - 9 | 1002 | 67 | |
XM_011537280.2 | XP_011535582.1 | 4 - 5 | 493 | 67 | |
XM_017021733.1 | XP_016877222.1 | 8 - 9 | 860 | 67 | |
XM_017021734.1 | XP_016877223.1 | 7 - 8 | 817 | 67 | |
XM_017021735.1 | XP_016877224.1 | 7 - 8 | 897 | 67 | |
XM_017021736.1 | XP_016877225.1 | 7 - 8 | 821 | 67 | |
XM_017021737.1 | XP_016877226.1 | 3 - 4 | 450 | 67 | |
XM_017021738.1 | XP_016877227.1 | 8 - 9 | 1982 | 67 | |
XM_017021739.1 | XP_016877228.1 | 8 - 9 | 2069 | 67 | |
GenBank mRNA | AB058727.1 | - | 7 - 8 | 1000 | 67 |
AF044774.1 | - | 2 - 3 | 163 | 67 | |
AL831932.2 | - | 7 - 8 | 918 | 67 | |
BC136632.1 | - | 7 - 8 | 1094 | 67 | |
BX648006.1 | - | 7 - 8 | 969 | 67 |