Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CD79A
Entrez Gene ID
973
Gene Name
CD79a molecule
Gene Aliases
IGA, MB-1
Chromosome Location
Chr.19: 41877120 - 41881372 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001783.3 NP_001774.1 2 - 3 561 88
GenBank mRNA AF283770.3 - 2 - 3 404 88
AK223371.1 - 2 - 3 559 88
AK315337.1 - 2 - 3 401 88
BC113731.1 - 2 - 3 396 88
BC113733.1 - 2 - 3 396 88
M74721.1 - 2 - 3 376 88
M80462.1 - 2 - 3 412 88
M86921.1 - 2 - 3 409 88
S46706.1 - 2 - 3 383 88
S75217.1 - 2 - 3 376 88

View Full Product Details